Druppelflessen, PP, Nalgene®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
DS2420-0125 DS2420-0250
216-4432EA 60.4 EUR
216-4432 NALGDS2420-0250
Druppelflessen, PP, Nalgene®
Flessen Druppelflessen
PPCO with non-drip closure and cap made from blue PP.
  • Autoclavable even when full with tips uncapped
  • Leakproof closure
  • Integral non-drip closure with attached cap
Bruikbaar voor steriele oplossingen.