Kopstukken voor gaswasflessen volgens Drechsel, Quickfit®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MF48/03/500 MF48/13/250 MF48/13/125 MF48/13/500 MF48/03/125 MF48/03/250 MF48/23/250 MF48/23/500 MF48/33/250
215-3398EA 73.7 EUR
215-3398 201-0245 201-0253 201-0254 215-3394 215-3395 BIBBMF48/23/250 215-3399 BIBBMF48/33/250
Kopstukken voor gaswasflessen volgens Drechsel, Quickfit®
Sluitingen Flesdoppen
PYREX®, borosilicate glass, with glass filter disc.
  • Porosity: 0/1/2 or 3
  • Very good chemical resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Sintered dip tube disperses gas into bubbles for greater efficiency.
BS 2461
Certificaten: BS 2461
Bestelinformatie: Drechsel bottle to be ordered separately.