Carboys, wide neck, HDPE, with screw cap
Leverancier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0431-1005 0431-1020 0431-1030 0431-1010 0431-0030 0431-0005 0431-0010 0431-0020
216-2852EA 44.5 EUR
216-2852 216-5710 216-2851 216-2850 216-5720 216-5717 216-5718 216-5719
Carboys, wide neck, HDPE, with screw cap
Flessen Voorraadflessen
HDPE, translucent.
  • Goede chemische bestendigheid
  • Met of zonder schroefdraadaansluiting voor het aansluiten van een tapkraan
Extra wide filling opening makes cleaning easier. Carrying handle and attached screw cap make it easy and safe to carry. The 5 and 10 litre sizes also have an air bleed valve. UV-stabilised.
Spigot or other accessories with ³/₄" inner thread can be attached to models with threaded connector.