Jerrycans, smalle hals, Nalgene®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2243-0013 2243-9013
216-4403EA 486 EUR
216-4403 216-4404
Jerrycans, smalle hals, Nalgene®
Flessen Jerrycans
Doorschijnend HDPE met PP-schroefdop.
  • Goede chemische bestendigheid
  • Temperatuurbestendig van −100 tot +120 °C
  • Zware, robuuste uitvoering, voldoet aan FDA 21CFR 177.1520 en USP-klasse VI
Ideal for storing reagents. The jerrican 216-4404 has two openings: a spout with attached screw cap (53 mm) and another (38 mm) with cap behind the carrying handle. Ideal as a reservoir in automatic systems such as the PRISM® 3700 DNA Analyser.
Order Now
Specificaties
Environmentally Preferable Lage impact op de fabricage
Milieu certificering NAR Certificate of Retirement