Jerrycans, zonder schroefdoppen
Leverancier: Kautex
Ratings:
Avg. Ratings: 4.0 (1 Review)
Total Ratings: 1
Avg. Ratings: 4.0 out of 5
Was this review helpful? Yes No
Yes: 1 No: 0 
Aéronautique / Approvisionneur / Villeurbanne
Yes, I recommend
Bonne résistance chimique Avec les bouchons adaptés aucun soucis d'étanchéité
353-80289 353-95642 353-86794 353-95982 353-95937 353-95939 353-95966 353-96232 353-96404 353-96138 353-92562 353-95981
216-5549EA 10.1 EUR
216-5549 216-5306 216-5551 216-4310 216-5548 216-5545 216-5352 216-4315 216-5302 216-1390 216-0415 216-1389
Jerrycans, zonder schroefdoppen
Flessen Jerrycans
HDPE, natural colour.
  • Jerrycans 2,5 to 5 L and 10 to 30 L with UN approval
  • Comprehensive range of closures
  • Varied use
  • Leakproof
  • Jerrycans are suitable for food
  • Halal certified
Universal jerrycan for industry, trade, and laboratories. These jerrycans, with a comprehensive range of closures accessories, can be used for almost any purpose.