VWR®, Mediaflessen met schroefdop
Leverancier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
215-2203EA 140 EUR
215-2203 215-2204 215-2205 215-2206 215-2210 215-2211 215-2924
VWR®, Mediaflessen met schroefdop
Flessen Mediumflessen
PC, autoclavable, narrow mouth bottles with standard PP caps.
  • Ideal for steam sterilisation
  • Safe and shatterproof
  • Chemically inert
VWR® media bottles are an ideal replacement for all borosilicate glass bottles. They are specifically designed for every laboratory application requiring terminal sterilisation (autoclaving), storage, transportation and production of laboratory biological fluids. These bottles are available in square and round shapes to accommodate various packaging needs.
The closure system is to be guaranteed leakproof, and the sealing ring allows convenient shrink-wrapping of the caps. Each bottle is carefully produced and quality controlled for uniform wall thickness, clarity, graduation and leakproof closure.
Optional septum cap or vented cap with PTFE membrane is also available.
Opgelet: *The product(s) on this page are outside the scope of the EU Directive 2019/904.