Flessen, smalle hals, rond, LDPE, met schroefdop, 301-serie
Leverancier: Kautex
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
301-770506 301-770502 301-770504 301-770503 301-770507 301-770500 301-770501 301-770505 301-770508 301-770509 301-770510 301-70511
215-9024EA 14.5 EUR
215-9024 215-9020 215-9022 215-9021 215-9025 215-9018 215-9019 215-9023 215-9026 215-9027 215-9028 215-9029
Flessen, smalle hals, rond, LDPE, met schroefdop, 301-serie
Flessen Kunststof flessen
LDPE. Round. Natural colour. With closure.
  • Flexible
  • Squeezable
  • Good shape-retaining properties
  • Halal certified
Suitable for food. Three types of closures for different applications. Ideal for liquids. Handy shape.