Flessen, smalle hals met schroefdoppen, Azlon®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BNP0125PN BNP0250P BNP0500P BNP1000P BNP2000P BNP0150PN
215-0529EA 26.9 EUR
215-0529 215-0530 215-0531 215-0532 215-0533 215-3109
Flessen, smalle hals met schroefdoppen, Azlon®
Flessen Kunststof flessen
PP, doorschijnend, met PP-schroefdop.
  • Very good chemical resistance
  • Autoclavable
  • Robust, leakproof
  • Heavy-duty and durable
Ideal for liquid storage, media preparation and sampling applications.
Certificaten: Suitable for food contact applications in accordance with EU and U.S. FDA regulations.