Flessen, smalle hals, rond, PE, met schroefdoppen
Leverancier: KARTELL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3171 3191 3231 3241 3251 3261
KART3171EA 3.3 EUR
KART3171 215-1358 215-1359 215-1360 215-1361 215-1362
Flessen, smalle hals, rond, PE, met schroefdoppen
Flessen Kunststof flessen
PE, grijs.
  • Double sealing with insert plug ensuring a leakproof seal
  • Easy grip cap
  • Opaque bottles ideal for the storage of photosensitive materials
Bottles have sloping shoulders reducing the tendency for residue deposits. Tagging points on the shoulders and caps to ensure tamper proof closure.