Flessen, type VERAL, smalle hals, met schroefdraad
Leverancier: MUELLER KREMPEL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
14.017.20 14.017.21 14.017.07 14.017.23
215-9151EA 0 EUR
215-9151 215-9152 215-3692 215-3694
Flessen, type VERAL, smalle hals, met schroefdraad
Flessen Glazen flessen
Clear glass or brown glass, without closure.
  • Good chemical and physical properties
  • For use in applications without high thermal loads
  • Opening PP 28 S, without pouring ring and screw cap (available separately)
Ideal for the sampling, storage and transportation of liquids. Brown-coloured bottles protect light-sensitive substances.
Order Now
Specificaties
Environmentally Preferable