Flessen, smalle hals, met standaard grondstoppen, PYREX®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1521/08D 1521/16D 1521/18D 1521/22D 1521/04D 1521/06D 1521/10D 1521/12D
215-0342EA 57.7 EUR
215-0342 215-0345 215-0346 215-0347 215-0340 215-0341 215-0343 BIBB1521/12D
Flessen, smalle hals, met standaard grondstoppen, PYREX®
Flessen Glazen flessen
PYREX®, borosilicate glass, with standard ground joint and glass stopper.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Met graduatie en wit schrijfveld.
ISO4796-2
Certificaten: ISO4796-2.
Order Now
Specificaties
Environmentally Preferable