Bottles, narrow or wide neck with standard ground joints
Leverancier: GRAVIS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M05254-30 M05364 M05372-12
215-2648EA 521 EUR
215-2648 215-2656 215-2658
Bottles, narrow or wide neck with standard ground joints
Flessen Glazen flessen
Soda-lime glass, clear or amber.
  • Good chemical and physical properties
  • Not thermal shock-resistant
  • Solid glass stoppers
Amber coloured to protect light-sensitive materials.