Flessen, smalle hals, rond, met schroefdraad
Leverancier: GRAVIS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A00112 A00212 A00218 A00118 A00102
215-2584EA 135 EUR
215-2584 215-2585 215-2586 216-0572 SPVAA00102
Flessen, smalle hals, rond, met schroefdraad
Flessen Glazen flessen
In soda-lime glass.
  • White (215-2583, 215-0572 and 215-2584)
  • Brown (215-2585 and 215-2586)
These screwed round mini bottles are available in three volumes.
Bestelinformatie: The capsules are ordered separately.