Flessen voor watermonsters, Sterilin®
Leverancier: STERLIN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
500WSC 500WSCNT WSC350 WSC350/NT
215-0385EA 426 EUR
215-0385 SLIN500WSCNT SLINWSC350 SLINWSC350/NT
Flessen voor watermonsters, Sterilin®
Flessen Watermonsternameflessen
Blow moulded from flexible polystyrene.
  • Sterile by gamma irradiation
  • Dosed bottle (20 mg/L) is suitable for neutralising samples of low chlorinated water
  • Tamper evident cap for sample integrity
  • Lot number for complete traceability
  • Closure material: HDPE
Convenient way of sampling both chlorinated and non-chlorinated water for microbiological analysis.
Available manufactured using:
Flexible polystyrene
NEW, lightweight PET for improved handling
Polystyrene bottles feature color-coded labels to aid differentiation:
Blue: dosed
Green: undosed
Available dose levels:
20 mg/h: Suitable for neutralizing water samples with a low concentration of chlorine
120 mg/h: Suitable for neutralizing water samples with a high concentration of chlorine