VWR®, Flessen, brede hals, met schroefdop
Leverancier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
215-5685EA 90.2 EUR
215-5685 215-5684 215-5683 215-5682 215-5681
VWR®, Flessen, brede hals, met schroefdop
Flessen Kunststof flessen
PP, doorschijnend.
  • Zeer goede chemische bestendigheid
  • Hittebestendig tot +121 °C (gedurende 20 minuten), langdurig gebruik tot +100 °C
  • Waterdicht
Perfect for sample collection, storage and transport. Ideal for liquids, pastes and granular material. The wide opening allows easy filling. Can be filled with hot substances. Come with easy to read blue graduations.