Flessen, brede hals met schroefdoppen
Leverancier: KARTELL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1608 1610 1612 1614 1616 1618
215-0139EA 3.5 EUR
215-0139 215-0140 215-0141 215-0142 215-0143 215-0144
Flessen, brede hals met schroefdoppen
Flessen Kunststof flessen
LDPE, translucent with LDPE screw cap.
  • Good chemical resistance
  • Heat resistant up to 75 °C
  • With graduations, leakproof
Multipurpose bottles for a wide variety of uses. Easy filling and dispensing of powders, pastes and liquids.
DIN 13316 and DIN 168
Certificaten: DIN 13316 and DIN 168