Flessen, brede hals met schroefdoppen, serie 303
Leverancier: Kautex
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
303-770531 303-770532 303-770533 303-770528 303-770529 303-770530 303-770534 303-770535 303-770536 303-770537 303-770528_SE 303-770529_SE 303-770531_SE 303-770533_SE 303-770535_SE 303-770537_SE
215-9037EA 16.6 EUR
215-9037 215-9038 215-9039 215-9034 215-9035 215-9036 215-9040 215-9041 215-9042 215-3113 KAUT303-770528_SE KAUT303-770529_SE KAUT303-770531_SE KAUT303-770533_SE KAUT303-770535_SE KAUT303-770537_SE
Flessen, brede hals met schroefdoppen, serie 303
Flessen Kunststof flessen
LDPE, round, natural colour, with closure.
  • Wide neck
  • Easy to fill
  • Easy to clean
  • Leakproof
  • Suitable for food
  • Halal certified
Ideal for liquids, pastes, and granulates.