Flessen, brede hals met schroefdoppen, serie 303
Leverancier: Kautex
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
303-783855 303-783856 303-783859 303-783852
215-9045EA 28.1 EUR
215-9045 215-9046 215-9047 215-9044
Flessen, brede hals met schroefdoppen, serie 303
Flessen Kunststof flessen
Polypropylene, round, natural colour.
  • Autoclavable up to 121 °C
  • Hot filled products, pastes, granulates
  • Leakproof
  • Halal certified
Suitable for food.
Note: Unscrew the cap and set on the bottle before autoclaving.
Caps are only suitable for bottles 215-9044 to 215-9047.