Flessen, brede hals, vierkant, met schroefdoppen, Azlon®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BGE212P BGE218P BGE224P BGE230P BGE336P BGE340P BGE342P
215-0438EA 43.1 EUR
215-0438 215-0440 215-0442 215-0444 215-0449 215-0451 215-0452
Flessen, brede hals, vierkant, met schroefdoppen, Azlon®
Flessen Kunststof flessen
HDPE, translucent.
  • Heavy-duty bottles with plug seal and screw cap
  • Easy to grip caps have a toggle for tamper-evident wire attachment to neck
  • Moulded graduations for approximate volume measurement, food safe
Order Now
Specificaties
Environmentally Preferable