Potten, brede hals met schroefdraad
Leverancier: Kautex
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
376-70541 376-70542 376-70543 376-70544 376-70545 376-71066 376-71492
215-2522EA 1.8 EUR
215-2522 215-3378 215-2523 215-3379 215-2524 215-2525 215-2521
Potten, brede hals met schroefdraad
Potjes Potjes voor algemeen gebruik
HDPE, round, natural colour.
  • Wide neck opening
  • Easy to fill and to clean
  • Leakproof
  • Suitable for food
  • Black closure with PE-foam wad is leakproof (closure to be ordered separately)
  • Halal certified
Due to the wide neck opening the containers are easy to fill and to clean. Particularly suitable for storage, transport, or disposal of powdery or granulated materials.
Leveringsinformatie:
The jars are supplied without lids, they can be ordered separately.