Opbergdozen voor objectglaasjes
Leverancier: BERGMANN KARTON
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
114 99 94
631-9011EA 26.4 EUR
631-9011 631-9012 631-9013
Opbergdozen voor objectglaasjes
Objectglaasjesdozen
These boxes are designed for 25, 50 and 100 slides.
  • Metal hinges and snap closure
  • Numbered racks with table of contents
  • Solid design with wooden frame and black coating