Transport- en opslagboxen
Leverancier: ZARGES LEICHTBAU
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
40861 40877 40862 40863
216-2121EA 0 EUR
216-2121 216-2122 216-2123 216-2124
Transport- en opslagboxen
Dozen Transportdozen
Aluminium alloy.
  • Lid with stainless steel strap hinge, two straps and all-round seal
  • Flip lock latches, secured with anti-tamper seals or padlock
  • Flip handles on the front surfaces
Secure transportation, practical storage and protective packaging in one. The transported or stored item is securely packed and protected against external influences. Sturdy edge, lid and base profile. Stackable.