Container for biohazardous waste, Jollypack
Leverancier: Allpack
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
LT20 LT40 LT60
129-1301EA 142 EUR
129-1301 129-1302 129-1303
Container for biohazardous waste, Jollypack
Dozen Afvaldozen
High quality corrugated cardboard with polyethylene bag.
  • Container have passed all the tests necessary for the required approval under regulations governing the collection and transportation of biohazardous hospital waste
  • They are collapsible to save storage space
  • Easy to assemble: Simply press two corners and the containers are ready to use
  • Each container comes with a polyethylene bag and a sealing strip
  • Container has temporary lid cover for daily use and permanent fixture for final secure sealing; lid stays on tightly even in the event of accidental