Emmers
Leverancier: Kautex
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
610-70589 615-70585 615B-70587
216-1536EA 47.2 EUR
216-1536 216-1534 216-1553
Emmers
Emmers
HDPE, with graduation.
  • Rustproof handle
  • Thick-walled
  • Sturdy
  • Suitable for food
  • Halal certified
Industrial and laboratory buckets are available with and without spout. Plastic coated steel handle. Builder’s buckets are dark grey with a rustproof handle. All buckets are thick-walled and very stable for a long working life and with graduated litre scale on the inside.