Burettes, straight stopcock, Schellbach stripe, PYREX®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
492092 492093 492094 492095 492145 492146 492147 492148
612-0214EA 58 EUR
612-0214 612-0215 612-0216 612-0217 612-0218 612-0219 612-0220 612-0221
Burettes, straight stopcock, Schellbach stripe, PYREX®
Buretten
Borosilicate glass, Class AS.
  • Calibrated to deliver ‘Ex’, conformity certified with dated batch identification
  • Funnel top to make filling easier and safer
  • Graduations and inscriptions in blue enamel
  • Choice of stopcock in glass or PTFE
DIN 12 700
Certificaten: ISO 385