Buretten, rechte kraan
Leverancier: HIRSCHMANN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3120160 3120170 3120175
612-3541EA 84.3 EUR
612-3541 612-3542 612-3543
Buretten, rechte kraan
Buretten
DURAN®, brown glass, class B.
  • Calibrated to deliver 'Ex'
  • Short marks at major graduations, white graduations
  • Glass key
Schaallengte: 500 mm
Certificaten: DIN EN ISO 385