Buretten, verwijderbare kraan
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BRB276P BRB284 BRB290 BRB264P BRB266P
BIBBBRB276PEA 45.4 EUR
BIBBBRB276P BIBBBRB284 612-0376 612-0369 612-0370
Buretten, verwijderbare kraan
Buretten
Soda lime glass, class B, removable PTFE stopcock.
  • Flared top for safer filling
  • Blue enamel graduations
BS 846
Certificaten: BS 846