Burettes, straight stopcock, PTFE spindle, MBL®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BRB402 BRB404 BRB406 BRB408
612-3254EA 83.4 EUR
612-3254 612-3256 612-3257 612-3258
Burettes, straight stopcock, PTFE spindle, MBL®
Buretten
Pyrex® borosilicate glass, class B.
  • Calibrated to deliver ‘Ex’
  • With GP Rotaflo®, stopcock which has a PTFE key, requiring no lubrication
  • Provides fine drop control, no seizure with caustic solutions
  • Graduations in permanent amber stain
Certificaten: ISO 385