Inoculating loops, Combo-loops
Leverancier: Copan
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
612-2674EA 163 EUR
612-2674 612-2675 612-2676 612-2677
Inoculating loops, Combo-loops
Uitstrijkspatels Entogen en -naalden
A range of nickel-chrome metal wire loops attached to lightweight insulated aluminium handles. Combo-loops are non calibrated and available in a range of loop sizes from 2 to 5 mm internal diameter. They offer the convenience of a ready to use loop and handle combination. Like regular nickel-chrome loops, Combo-Loops can be sterilised between use by using an incinerator or Bunsen flame. The whole unit is designed to be discarded when the wire loop eventually wears out.
  • Combined loop plus handle
  • Combo-Loop handles are colour coded to facilitate easy size identification
  • Loops have totally smooth surfaces and will not cut or gouge through agar medium when streaking plate cultures
Verpakking: Loops are supplied in packages containing five loops with handles.