Naalden
Leverancier: MEDICAL WIRE EQUIPMENT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MW702/1 MW702/20
632-0056EA 299 EUR
632-0056 632-0057
Naalden
Uitstrijkspatels Entogen en -naalden
PS, sterile. Disposable plastic inoculating needles.
  • Moderately flexible
  • Colour coded plastic