Klemmen voor conische slijpingen
Leverancier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
01103712 01103719 01103724 01103734 01103745 01103714 01103729
201-0765EA 37.6 EUR
201-0765 271-1121 271-1122 271-1124 201-0767 201-0900 201-0901
Klemmen voor conische slijpingen
Klemmen NS-verbindingsklemmen
POM, coloured. For firm, securely fitting conical ground joints. These clamps are made of plastic to avoid scratching the glass, as happens with conventional metal joint clamps.
  • Very good chemical resistance
  • Heat resistant up to +150 °C
  • Sturdy, easy to install and remove