Tubing clamps
Leverancier: BOSCH BERND
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SK-10 SK-20
229-0351EA 0 EUR
229-0351 229-0353
Tubing clamps
Klemmen Slangklemmen
Hooks and sliding clamps in galvanized steel, locking nut in plastic.
  • Heat resistant up to 150 °C
  • Lightweight, suitable for thick-walled vacuum and pressure hoses
  • No need to tip the clamping slide, so no accidental slipping or opening of the hose clamp when it's closed
The plastic locking nut (229-0351 with aluminium locking nut) allows flow control or a total stop of the flow, even at already existing hose connections. The closure of the clamp can be opened in any position without screwing and unscrewing.