Artery forceps, Kocher (Ochsner) and Pean type
Leverancier: HAMMACHER KARL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HSE066-16 HSE088-16 HSE072-20 HSE095-20 HSE089-16 HSE092-18 HSE062-13 HSE065-14 HSE067-16
HAMMHSE066-16EA 111 EUR
HAMMHSE066-16 221-0056 232-4162 HAMMHSE095-20 221-0057 HAMMHSE092-18 HAMMHSE062-13 HAMMHSE065-14 HAMMHSE067-16
Artery forceps, Kocher (Ochsner) and Pean type
Pincetten
Stainless steel. These forceps are available in straight or curved format.
  • Anti-magnetic
  • Haemostatic, high medical quality
  • Pean or Kocher version