Bottles, narrow neck, square, HDPE, UN approved with thread
Leverancier: Kautex
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
308-770096 310-84486 310-770097 310-84152 310-84153 308-84464
215-0098EA 395 EUR
215-0098 215-3949 215-3945 215-3946 215-3947 215-3948
Bottles, narrow neck, square, HDPE, UN approved with thread
Flessen Kunststof flessen
HDPE, naturel of zwart.
  • UN approval
  • For hazardous liquids
  • Three different types of closures
  • GL 45 neck
  • Leakproof
  • Bottles are suitable for food
  • Recessed labelling field with bottle sizes 1000 and 2500 ml
  • Halal certified
UN approved for liquids. Bottle includes a dust cap to avoid contamination before filling. Dispose of dust cap prior to capping.