Bottles, narrow neck, with screw cap
Leverancier: GLASWARENFABRIK KARL HECHT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
41322030 41323072 41322012 41323054 41322066 41322054 41322018 41322072
215-2194EA 106 EUR
215-2194 HECH41323072 HECH41322012 HECH41323054 HECH41322066 HECH41322054 HECH41322018 HECH41322072
Bottles, narrow neck, with screw cap
Flessen Glazen flessen
Narrow neck bottles, clear or brown glass, with black PP screw cap. Excellent for sampling, storing and transporting liquids.
  • Good chemical and physical properties
  • For applications that do not involve high thermal stresses
  • Round thread in accordance with DIN 168
  • Brown coloured for light sensitive substances