Bottles, round with wide neck or flat with narrow neck
Leverancier: GRAVIS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
B00347 B00364 D00747PP28 B00354EURO
215-2588EA 115 EUR
215-2588 215-2590 215-2621 SPVAB00354EURO
Bottles, round with wide neck or flat with narrow neck
Flessen Glazen flessen
Clear glass.
  • 2 types of stoppers: self-sealing PE or bakelite with rubber seal (sterilisable at +120 °C)
Bestelinformatie: Bottles provided without cap (order separately).