Flessen, brede hals, rond, met schroefdraad
Leverancier: GRAVIS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
H02758 H02770 H02768
215-2641EA 28.3 EUR
215-2641 215-2418 215-3235
Flessen, brede hals, rond, met schroefdraad
Flessen Glazen flessen
Clear glass.
  • Very wide aperture, screw ring
  • Metal lids, molded joint, resistant up to +120 °C
  • Different formats available
Bestelinformatie: Lids must be ordered separately.