Dropper caps for narrow neck bottles, 301 and 302 series
Leverancier: Kautex
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
406-72103 419-73350 455-72100 455-72101 455-72102
215-6320EA 0.6 EUR
215-6320 215-6301 215-0920 215-1409 215-6305
Dropper caps for narrow neck bottles, 301 and 302 series
Sluitingen Druppeldoppen
LDPE, natural colour.
  • Good chemical resistance
  • Good thermal resistance
Can only be used with 301 and 302 series narrow mouth bottles.
Bestelinformatie: The red PVC captive caps slide on tapered closures of varying sizes.