Drip closures
Leverancier: KARTELL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3303 3304 3305 3306 3307 3308
217-0488EA 2.4 EUR
217-0488 217-0489 217-0490 217-0491 217-0492 KART3308
Drip closures
Sluitingen Druppeldoppen
HDPE, natural colour.
  • Good chemical resistance, good temperature resistance
  • Available in two lengths
  • Attached cap
For narrow-neck bottles with round thread in accordance with DIN 168.