Anion exchange chiral stationary phases
Leverancier: CHIRAL TECHNOLOGIES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
32324 31311 31324 31394 32311 32394 31344 32344
554-3754EA 1900 EUR
554-3754 554-3750 554-3765 554-3752 554-3753 554-3756 554-3751 554-3755
Anion exchange chiral stationary phases
Kolommen voor chromatografie
For the separation of acidic chiral compounds containing carboxylic, phosphonic, phosphinic, phosphoric or sulphonic acid groups. In some cases weakly acidic compounds, such as phenols, can also be separated.