Fractionerings kolommen
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
FC1/12 FC1/22
201-8387EA 157 EUR
201-8387 201-8388
Fractionerings kolommen
Onderdelen voor destillatie Destillatiekolommen
PYREX®, borosilicaatglas
  • Very good chemical resistance
  • High temperature resistance
Comprises column with accurately fitted Dufton glass spiral. Good efficiency - liquid spiralling down column meets ascending vapour. Spiral can be removed for cleaning.