Ferrules voor capillaire kolommen, GVF
Leverancier: Trajan Scientific and Medical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
072651 072652 072653 0726560 0726591 0726640
558-0191EA 64.8 EUR
558-0191 558-0192 558-0194 568-0473 568-0474 558-0209
Ferrules voor capillaire kolommen, GVF
Fittingen en connectoren
These ferrules are made from 15% graphite and 85% Vespel®. They have a temperature limit of 350 °C and cannot be re-used with another capillary column.
  • Mechanically robust, hard material, long lifetime
  • Forms strong grip with column
  • Must be re-tightened after initial temperature cycles
  • For GC-MS interface
Poorly defined or selected connections can lead to an increase in dead volume, leaks from cycled temperatures and mismatched tubing sizes. Selecting the correct GC connections will improve your chromatography.
Temperature limit 350 °C.
Cannot be re-used.