HPLC columns, Waters Spherisorb®
Leverancier: WATERS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PSS837515 PSS832715 PSS837513 PSS832713 PSS831013 PSS831012 PSS831812 PSS830612 PSS832514 PSS832213 PSS833413 PSS832212 PSS832211 PSS833411 PSS832111 PSS839510 PSS831915 PSS832515 PSS831913 PSS831912 PSS832513 PSS830915 PSS830715 PSS830713 PSS830613 PSS830614 PSS830714 PSS833412 PSS832112 PSS830113 PSS830115 PSS832613 PSS832615 PSS831015 PSS831815 PSS831813 PSS830913 PSS831113 PSS831115 PSS830616 PSS830615 PSS831914 PSS830813 PSS830815 PSS832113 PSS830909 PSS845277 PSS845542
554-0987EA 0 EUR
554-0987 554-0982 554-0986 554-0981 554-0959 554-0958 554-0963 554-0946 554-0977 554-0975 554-0985 554-0974 554-0973 554-0983 554-0970 554-0991 554-0969 554-0978 554-0967 554-0966 554-0976 554-0957 554-0953 554-0951 554-0947 554-0948 554-0952 554-0984 554-0971 554-0944 554-0945 554-0979 554-0980 554-0960 554-0965 554-0964 554-0956 554-0961 554-0962 554-0950 554-0949 554-0968 554-0954 554-0955 554-0972 WATEPSS830909 WATEPSS845277 WATEPSS845542
HPLC columns, Waters Spherisorb®
Kolommen voor chromatografie
Spherisorb® is a porous, spherical material with a range of narrow particle and pore size distribution resulting in very efficient chromatographic separations. These columns have industry standard, fixed end-fittings and are available in three particle sizes. For guard columns, use corresponding Spherisorb® guard cartridge with stand-alone holder listed under “Fittings and Accessories'”
  • Wide range of sizes and bonded phases
  • Silica is produced in 3, 5, and 10 µm particle sizes