Tweedelige verbindingsstukken
Leverancier: VICI JOUR RESEARCH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
JR-M5570 JR-M5003
554-1155EA 118 EUR
554-1155 554-1066
Tweedelige verbindingsstukken
Fittingen en connectoren
PEEK two-piece fingertight fittings. Fits any 10 to 32 fitting. Works with any ¹/₁₆'' OD tubing.
  • Nominale druk 280 bar (4000 psi)
  • Handvast
  • Herbruikbare moer, vervangbare eindhuls