Finger tight fittings, geflensd
Leverancier: VICI JOUR RESEARCH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
JRM201094BLACK JR-201926-10 JR-201928-10
554-1139EA 22.2 EUR
554-1139 554-0750 554-1141
Finger tight fittings, geflensd
Fittingen en connectoren
Fiberglass reinforced PP with PP washers. A number of chromatography systems still require or prefer flanged connections. When the tubing is flanged, the washer never comes in contact with the mobile phase.
  • Compatible with all 1/4"-28 flat-bottom ports
  • Available for 1/16" and 1/8" tubing
Pressure rating: Up to 30 bar (435 psi).
Pressure rating: Up to 30 bar (435 psi).