Eendelige zeshoekige fitting
Leverancier: VICI Jour
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
554-1153EA 100 EUR
554-1153 554-1154
Eendelige zeshoekige fitting
Fittingen en connectoren
Peek one-piece hex-head fittings, with their integrated ferrule, are easy to use with the valve-tool and the colour-it colour-coded finger-tight adapters. PEEK one-piece hex-head fittings are available in short and long versions.
Peek one-piece hex-head fittings, with their integrated ferrule, are easy to use with the valve-tool and the colour-it colour-coded finger-tight adapters. PEEK one-piece hex-head fittings are available in short and long versions.
  • Ideal for use with PEEK tubing
  • 10-32 thread will fit virtually all standard high-pressure female connection
Withstand pressures up to 350 bar (5000 psi)
  • Ideal for use with PEEK tubing
  • 10-32 thread will fit virtually all standard high-pressure female connection
Withstand pressures up to 350 bar (5000 psi)