VWR® Hichrom Column End Plugs
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HI-001EA 17.8 EUR
HI-001 HICHHI-001K
VWR® Hichrom Column End Plugs
Fittingen en connectoren
These polymer column end plugs are used to prevent solvent evaporation when the columns are not in use.
  • Fingertight design
  • Extends column life
  • Fits any size columns