VWR® Hichrom Cutters, Polymeric Tubing
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
554-7743EA 48.9 EUR
554-7743 554-7745
VWR® Hichrom Cutters, Polymeric Tubing
Snijders
The Clean-cut™ cutter is designed specifically for cutting polymeric HPLC tubing from 1/16” to 1/8” with no distortion of cross-section and without closing the bore.
  • Cuts PEEK and PTFE tubing
  • Square, burr-free cuts
  • Convenient, replaceable blade
The “Guillotine” cutter is an alternative design for cutting polymeric tubing of 1/16” to 1/2”. A razor-sharp blade cuts while a V-shaped grove on the handle holds the tubing at the right angle to produce a square cut.