Last drop filter/ontgassing
Leverancier: VICI JOUR RESEARCH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
JR-9000-0602 JR-9000-0603 JR-9000-0604 JR-9000-0640 JR-9000-0641 JR-9000-0642
554-1118EA 232 EUR
554-1118 554-1119 554-1120 554-1121 554-1122 554-1123
Last drop filter/ontgassing
Filters Filters voor vloeistofverwerking
The Last Drop filter/sparger combines filtration and sparging in a single unit. The PTFE housing contains a mobile phase filter with either a stainless steel or PTFE filter element. Connect the solvent line to the stepped connector, which accommodates 1,5, 2,2, and 3,5 mm ID tubing. A 1/4"-28 fitting connector connects a 1/8" helium line connects to the 10 µm sparger. Both connectors are PEEK.
  • Parallel filtering and sparging
  • Biocompatible PTFE or SS frits