Spiraalkoelers met NS hals, Quickfit®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CX3/05/SC CX3/06/SC
201-8300EA 167 EUR
201-8300 201-8301
Spiraalkoelers met NS hals, Quickfit®
Condensors
PYREX®, borosilicaatglas.
  • Highly efficient condensers with a large cooling surface
  • Very good chemical and temperature resistance
Coil type, with ground cone. Ideal for use in the upright reflux mode where counter-current cooling is possible. Detachable plastic connectors for the attachment of rubber hose.
ISO 1773, BS 6352
Certificaten: ISO 1773, BS 6352